missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nickel(II) fluoride tetrahydrate, Puratronic™, 99.99% (metals basis)

Brand:  Alfa Aesar™ 010817.04

UNSPSC : 12141729

Code : NEW

Additional Details : CAS Number : 13940-83-5 Weight : 0.00200kg Transport : UN number : 3288 Chem class : 1 Pack group : III

Quantity 2g

 View more versions of this product

Product Code. 11306947

  • £29.57 / 2g


Estimated Shipment date
from Supplier 18-10-2017
Add to basket

Specifications

Chemical Identifiers

Nickel(II) fluoride tetrahydrate
Puratronic™
168.77 (96.71 Anhydrous)
UN3288
13940-83-5
NiF2·4H2O
MFCD00016263

Specifications

Nickel(II) fluoride tetrahydrate
Puratronic™
99.99%
NiF2·4H2O
MFCD00016263
4.6g/mL
Green
13940-83-5
Powder
(metals basis)
168.77 (96.71 Anhydrous)
UN3288
Odorless
2g
Documentation

Technical Documents