missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) fluoride tetrahydrate, 98+%, Thermo Scientific Chemicals
25.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
Description
Nickel(II) fluoride tetrahydrate is used in surface treatment of aluminum profile and printing ink making, fluorescence lamp, fluidizer and catalyzer for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 50 g |
UN Number | UN3288 |
Solubility Information | Soluble in water,inorganic acid. |
Formula Weight | 168.77 (96.71 Anhydrous) |
Percent Purity | ≥98% |
Odor | Odorless |
Chemical Name or Material | Nickel(II) fluoride tetrahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'