missing translation for 'onlineSavingsMsg'
Learn More

Nickel(Ii) Chloride Hexahydrate, ReagentPlus™ Honeywell™

Product Code. p-7120618 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
25 g
500 g
2 kg
Packaging:
Poly Bottle
Unit Size:
25g
2kg
500g
This item is not returnable. View return policy

Product Code. 15632200

Brand: Honeywell Research Chemicals 22338725G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-20-0
MDL Number MFCD00149809

Specifications

Quantity 25 g
Linear Formula NiCl2 · 6H2O
UN Number UN3288
Formula Weight 237.69g/mol
Grade ReagentPlus™
Packaging Poly Bottle
Chemical Name or Material Nickel(Ii) Chloride Hexahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.