missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) acetate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog Number 11974901
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
6018-89-9
C4H14NiO8
248.84
MFCD00066973
OINIXPNQKAZCRL-UHFFFAOYSA-L
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
62601
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O
This item is not returnable. View return policy
6018-89-9
C4H14NiO8
248.84
MFCD00066973
OINIXPNQKAZCRL-UHFFFAOYSA-L
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
62601
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 250.0°C
Color White
Physical Form Crystalline Powder, Crystals or Flakes
Quantity 2.5 kg
Assay Percent Range 99% min. (Complexometry)
Linear Formula (CH3CO2)2Ni·4H2O
Fieser 14,288; 16,308
Merck Index 14,6498
Solubility Information Solubility in water: 182g/L (20°C). Other solubilities: soluble in alcohol
IUPAC Name nickel(2+) diacetate tetrahydrate
Formula Weight 248.86
Percent Purity ≥99%
Grade Analytical
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Nickel(II) acetate tetrahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'