missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) 2-ethylhexanoate monoisopropoxide, 99+% (m.b.), 5% w/v in IPA, pkgd under Ar in rslble ChemSeal bottles, Thermo Scientific™
Click to view available options
Quantity:
25 mL
100 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged under Argon
Specifications
Packaging | ChemSeal™ bottles |
Quantity | 100 mL |
UN Number | UN1219 |
Sensitivity | Moisture sensitive |
Formula Weight | 261 |
Percent Purity | ≥99% (m.b.) |
Concentration or Composition (by Analyte or Components) | 5% w/v in isopropanol |
Physical Form | Liquid |
Chemical Name or Material | Nickel (II) 2-ethylhexanoate monoisopropoxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'