Learn More
Alfa Aesar™ Nickel Cover for Crucible
Brand:  Alfa Aesar™ 036094.KT
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15441437
  • £9.75 / Each
Estimated Shipment date
from Supplier 01-02-2018
Add to basket
Specifications
Specifications
20mL
For 35952 crucible
Nickel Crucible Cover
Nickel