missing translation for 'onlineSavingsMsg'
Learn More

Nickel Copper foil, alloy 400, 1.6mm (0.062 in.) thick, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 045231.GT

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15416845

  • £179.00 / Each
Estimated Shipment: 20-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Nickel Copper foil, Alloy 400
Ni:Cu; 67:33 wt%
150 x 150 mm
1.6mm (0.062 in.) thick
MFCD00801098
Foil
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers