missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Nickel Copper foil, alloy 400, 1.0mm (0.04 in.) thick

Brand:  Thermo Scientific Alfa Aesar 045225.GT

Nickel Copper foil, alloy 400, 1.0mm (0.04 in.) thick

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15476835

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.