missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nickel Copper foil, alloy 400, 1.0mm (0.04 in.) thick

Brand:  Alfa Aesar™ 045225.HB

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300x300mm

 View more versions of this product

Product Code. 15486835

  • £279.10 / Each


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

Nickel Copper foil, Alloy 400
1.0mm (0.04 in.) thick
MFCD00801098

Specifications

Ni:Cu; 67:33 wt%
300x300mm
Foil
Nickel Copper foil, Alloy 400
1.0mm (0.04 in.) thick
MFCD00801098
Videos
SDS