missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Nickel Chromium Iron rod, 6.4mm (0.25 in.), hot finished, annealed

Brand:  Thermo Scientific Alfa Aesar 036455.G1

Nickel Chromium Iron rod, 6.4mm (0.25 in.), hot finished, annealed

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11327918

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.