missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chromium gauze, Alfa Aesar™

Open area: 36%; Width of opening: 0.015 in.

Brand:  Alfa Aesar 040941.HB

Code : NEW

Additional Details : CAS Number : 11106-97-1 Weight : 0.01000kg

Quantity 300 x 300mm

 View more versions of this product

Product Code. 11375257

  • £658.60 / Each


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Nickel Chromium
11106-97-1
Ni:Cr; 80:20 wt%
gauze
Wire Cloth
MFCD00673110

Specifications

Nickel Chromium
300 x 300mm
Silver -colored gauze
Odorless
Ni:Cr; 80:20 wt%
Silver
gauze
11106-97-1
Wire Cloth
MFCD00673110
Videos
SDS