Learn More
Nickel Chromium gauze, Alfa Aesar™
Open area: 36%; Width of opening: 0.015 in.
Brand: Alfa Aesar 040941.FL
Code : NEW
Additional Details : CAS Number : 11106-97-1 Weight : 0.01000kg
Quantity | 75 x 75mm |
---|
Chemical Identifiers
Nickel Chromium | |
11106-97-1 | |
Ni:Cr; 80:20 wt% |
gauze | |
Wire Cloth | |
MFCD00673110 |
Specifications
Nickel Chromium | |
75 x 75mm | |
Silver -colored gauze | |
Odorless | |
Ni:Cr; 80:20 wt% |
Silver | |
gauze | |
11106-97-1 | |
Wire Cloth | |
MFCD00673110 |