missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel chromite, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
In mixing Nickel (II) Chromate with Aluminates or Cobalt Chromate, a binary compound known as a spinel is formed that is able to withstand temperatures up to 1000 °C. The products from these mixings can be used as oxidation-resistive components in airplane and space shuttle coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 100 g |
Linear Formula | NiCr2O4 |
Solubility Information | Insoluble in water. Soluble in hydrochloric acid. |
Formula Weight | 226.7 |
Odor | Odorless |
Chemical Name or Material | Nickel chromite |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'