missing translation for 'onlineSavingsMsg'
Learn More

Nickel boride, 99%, Alfa Aesar™

Brand:  Alfa Aesar 013109.14

Code : NEW

Additional Details : CAS Number : 12007-01-1 Weight : 0.02500kg Transport : UN number : 3077 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 11372198

  • £54.10 / 25g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Nickel boride
-35 Mesh Granular
Ni2B
MFCD00084757
12007-01-1
99%
128.23

Specifications

Nickel boride
-35 Mesh Granular
Ni2B
MFCD00084757
12007-01-1
99%
128.23
25g
Videos
SDS