Learn More
Alfa Aesar™ Nickel based steam reforming catalyst, HiFUEL™ R110
Brand:  Alfa Aesar™ 045465.A1
Code : NEW
Additional Details : Weight : 1.00000kg
Quantity 1kg
 View more versions of this product
Product Code. 15406935
  • £195.36 / 1kg
Estimated Shipment date
from Supplier 10-05-2018
Add to basket
Specifications
Chemical Identifiers
Nickel based steam reforming catalyst HiFUEL™ R110
Specifications
Nickel based steam reforming catalyst HiFUEL™ R110
Pale gray to green
Odorless
4-Hole, 4-Flute Domed Cylinders
1kg