missing translation for 'onlineSavingsMsg'
Learn More

Nickel Aluminum, Raney|r type non-activated, Thermo Scientific Chemicals

Catalog Number 11395009
Click to view available options
Quantity:
50 g
500 g
2 kg
This item is not returnable. View return policy
12635-29-9
AlNi
85.675
MFCD00167392
NPXOKRUENSOPAO-UHFFFAOYSA-N
aluminum-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminum nickel
6336846
aluminum;nickel
[Al].[Ni]
This item is not returnable. View return policy
12635-29-9
AlNi
85.675
MFCD00167392
NPXOKRUENSOPAO-UHFFFAOYSA-N
aluminum-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminum nickel
6336846
aluminum;nickel
[Al].[Ni]

Nickel Aluminum is used for the manufacture of aluminum-nickel alloy powder, and it is used in the chemical, pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black to Gray
Avg. Mol. Wt. or Mol. Wt. Range Ni:Al
UN Number UN3208
Quantity 50 g
Solubility Information Insoluble in water.
Odor Odorless
Sensitivity Water reactive
Form Powder
Chemical Name or Material Nickel Aluminum, Raney™ type non-activated

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'