missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Aluminum, Raney|r type non-activated, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
500 g
2 kg
Description
Nickel Aluminum is used for the manufacture of aluminum-nickel alloy powder, and it is used in the chemical, pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black to Gray |
Avg. Mol. Wt. or Mol. Wt. Range | Ni:Al |
UN Number | UN3208 |
Quantity | 50 g |
Solubility Information | Insoluble in water. |
Odor | Odorless |
Sensitivity | Water reactive |
Form | Powder |
Chemical Name or Material | Nickel Aluminum, Raney™ type non-activated |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'