missing translation for 'onlineSavingsMsg'
Learn More

Nickel aluminum, 99.0% min (metals basis), Thermo Scientific Chemicals

Catalog Number 11318117
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
12003-78-0
AlNi
85.675
MFCD00167392
NPXOKRUENSOPAO-UHFFFAOYSA-N
aluminum-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminum nickel
6336846
aluminum;nickel
[Al].[Ni]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
12003-78-0
AlNi
85.675
MFCD00167392
NPXOKRUENSOPAO-UHFFFAOYSA-N
aluminum-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminum nickel
6336846
aluminum;nickel
[Al].[Ni]

Nickel aluminum is used in strengthening constituent in high-temperature nickel-base superalloys, coating blades in gas turbines and jet engines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Appearance Gray powder, Intermetallic
Assay Percent Range 99.0% min. (metals basis)
Quantity 250 g
Solubility Information Insoluble in water.
Odor Odorless
Form Powder
Chemical Name or Material Nickel aluminum

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'