missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nickel 61, 61Ni, plasma standard solution, Specpure™, 61Ni 10μg/mL

Brand:  Alfa Aesar™ 045282.AD

Code : NEW

Additional Details : Weight : 0.00050kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Quantity 50mL

Product Code. 15456875

  • £226.60 / 50mL


Estimated Shipment date
from Supplier 22-05-2018
Add to basket

Specifications

Specifications

Nickel 61, 61Ni, plasma standard solution
Specpure™
MFCD00144901
50mL
61Ni 10μg/mL
Liquid
UN3264
Matrix: 2% HNO3
Videos
SDS