missing translation for 'onlineSavingsMsg'
Learn More

NGD 94-1, Tocris Bioscience™

Selective D4 receptor ligand

Brand:  Tocris (Bio-Techne) 329810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13462734

  • £124.20 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

High affinity D4 receptor ligand; selective over D1, D2, D3 and D5 receptors. Displays antagonist activity at the human D4.2 receptor (Ki = 3.6 nM in transfected CHO cells) and exhibits agonist activity at the D4.4 receptor in HEK 293 cells.
Specifications

Chemical Identifiers

178928-68-2
320.4
unii-do9m06bfrj, do9m06bfrj, 2-4-2-phenyl-1h-imidazol-4-yl methyl piperazin-1-yl pyrimidine, 2-4-2-phenyl-1h-imidazol-5-yl methyl piperazin-1-yl pyrimidine, 2-phenyl-4 5-4-2-pyrimidinyl-piperazin-1-yl methyl imidazole dimaleate, 2-4-2-phenyl-1h-imidazol-5-yl methyl-1-piperazinyl-pyrimidine, 2-4-2-phenyl-3h-imidazol-4-yl methyl piperazin-1-yl pyrimidine, 2-4-2-phenyl-3h-imidazol-4-yl methyl piperazin-1-yl pyrimidine, d0f7nm, d0l3ji
2-[4-[(2-phenyl-1H-imidazol-5-yl)methyl]piperazin-1-yl]pyrimidine
C18H20N6
OTVQCHUIPJYASM-UHFFFAOYSA-N
188942
C1CN(CCN1CC2=CN=C(N2)C3=CC=CC=C3)C4=NC=CC=N4

Specifications

NGD 94-1
178928-68-2
unii-do9m06bfrj, do9m06bfrj, 2-4-2-phenyl-1h-imidazol-4-yl methyl piperazin-1-yl pyrimidine, 2-4-2-phenyl-1h-imidazol-5-yl methyl piperazin-1-yl pyrimidine, 2-phenyl-4 5-4-2-pyrimidinyl-piperazin-1-yl methyl imidazole dimaleate, 2-4-2-phenyl-1h-imidazol-5-yl methyl-1-piperazinyl-pyrimidine, 2-4-2-phenyl-3h-imidazol-4-yl methyl piperazin-1-yl pyrimidine, 2-4-2-phenyl-3h-imidazol-4-yl methyl piperazin-1-yl pyrimidine, d0f7nm, d0l3ji
C1CN(CCN1CC2=CN=C(N2)C3=CC=CC=C3)C4=NC=CC=N4
320.4
320.39
10mg
C18H20N6
OTVQCHUIPJYASM-UHFFFAOYSA-N
2-[4-[(2-phenyl-1H-imidazol-5-yl)methyl]piperazin-1-yl]pyrimidine
188942
>99%
Videos