missing translation for 'onlineSavingsMsg'
Learn More

Ngamma-Trityl-L-asparagine hydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-7054860
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
132388-58-0
C23H22N2O3
374.44
MFCD00153422
BRRPJQYCERAMFI-GNLPSFAGNA-N
h-asn trt-oh, n'-trityl-l-asparagine, n-beta-trityl-l-asparagine, n-triphenylmethyl-l-asparagine, 2s-2-amino-4-oxo-4-tritylamino butanoic acid, 2s-2-amino-3-triphenylmethylcarbamoyl propanoic acid, 2s-2-amino-3-triphenylmethyl carbamoyl propanoic acid, h-asn trt-2-chlorotrityl resin, asparagine trt, h-asn trt oh
7408309
(2S)-2-amino-4-oxo-4-(tritylamino)butanoic acid
N[C@@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
This item is not returnable. View return policy
132388-58-0
C23H22N2O3
374.44
MFCD00153422
BRRPJQYCERAMFI-GNLPSFAGNA-N
h-asn trt-oh, n'-trityl-l-asparagine, n-beta-trityl-l-asparagine, n-triphenylmethyl-l-asparagine, 2s-2-amino-4-oxo-4-tritylamino butanoic acid, 2s-2-amino-3-triphenylmethylcarbamoyl propanoic acid, 2s-2-amino-3-triphenylmethyl carbamoyl propanoic acid, h-asn trt-2-chlorotrityl resin, asparagine trt, h-asn trt oh
7408309
(2S)-2-amino-4-oxo-4-(tritylamino)butanoic acid
N[C@@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Formula Weight 392.45 (Anhydrous)
Percent Purity 98%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material Ngamma-Trityl-L-asparagine hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'