Learn More
Invitrogen™ Human NGAL, Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP75672
Code : LD
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
NGAL (human) Recombinant Protein, RP-75672, from Invitrogen.
NGAL (Neutrophil gelatinase-associated lipocalin, lipocalin 2, siderocalin and neutrophil lipocalin) is a 25 kDa glycosylated single chain monomer and member of the lipocalin family of proteins that bind and transport small lipophilic molecules. NGAL is released by activated neutrophils, can form dimers and small amounts of higher oligomers, and complexes with matrix metalloproteinase 9 (MMP-9, gelatinase B). Low level expression of NGAL in a variety of epithelia may be increased in inflammation or cancers. Expression of NGAL in the kidney is dramatically increased by ischemic or nephrotoxic kidney injury. This product contains purified recombinant human NGAL protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays. This protein has a molecular weight of ∽27.1 kDa.Specifications
P80188 | |
Control, Western Blot | |
3934 | |
7.4 | |
E. Coli | |
RUO | |
Human NGAL | |
Human | |
Hu | |
Recombinant | |
Recombinant |
0.43 mg/mL | |
PBS with 50% glycerol and no preservative; pH 7.4 | |
Human NGAL | |
100μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
Neurophil Gelatinase Associated Lipocalin | |
LCN2 | |
Unconjugated | |
E. coli | |
Liquid | |
purified |