Learn More
NFKBIL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555153
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: WNRRNDMGET LLHRACIEGQ LRRVQDLVRQ GHPLNPRDYC GWTPLHEACN YGHLEIVRFL LDHGAAVDDP GGQGCEGITP LHDALNCGHF EVAELLLERG ASVTLRTRK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
NFKBIL2 Polyclonal antibody specifically detects NFKBIL2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
NFKBIL2 | |
Polyclonal | |
Unconjugated | |
TONSL | |
DNA repair protein; ikappaBR; I-kappa-B-related protein; IKBR; Inhibitor of kappa B-related protein; NF-kappa-B inhibitor-like protein 2; NFKBIL2; Nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 2; TONSL; tonsoku-like; Tonsoku-like protein; tonsoku-like, DNA repair protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4796 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96HA7 | |
TONSL | |
Recombinant protein corresponding to Human NFKBIL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only