missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NF-M Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | NF-M |
---|---|
Clone | 327921 |
Concentration | LYOPH |
Dilution | Western Blot 1-3 ug/mL, Knockout Validated |
Applications | Western Blot, KnockDown |
Description
NF-M Monoclonal specifically detects NF-M in Human samples. It is validated for Western Blot, Knockout Validated.Specifications
NF-M | |
LYOPH | |
Western Blot, KnockDown | |
Unconjugated | |
Mouse | |
Human | |
P07197 | |
4741 | |
E. coli-derived recombinant human NF-M Ser2-Ser358 Accession # P07197 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human NF-M in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) NF-H or rhNF-L is observed. |
327921 | |
Western Blot 1-3 ug/mL, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
NEF3, NEF3neurofilament medium polypeptide, NEFM, Neurofilament 3, neurofilament, medium polypeptide, neurofilament, medium polypeptide 150kDa, neurofilament-3 (150 kD medium), NFM, NF-M160 kDa neurofilament protein, NFMNeurofilament triplet M protein | |
NEFM | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only