missing translation for 'onlineSavingsMsg'
Learn More

NF 279, Tocris Bioscience™

Potent and selective P2X1 antagonist

Brand:  Tocris (Bio-Techne) 119950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13488843

  • £625.89 / 50mg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

A potent and selective P2X1 antagonist (IC50 = 19 nM). Displays good selectivity over P2X2,(IC50 = 0.76 μM), P2X3 (IC50 = 1.62μM), P2X4 (IC50 > 300 μM), P2Y receptors and ecto-nucleotidases.
Specifications

Chemical Identifiers

202983-32-2
1407.145
8,8'-[Carbonylbis(imino-4,1-phenylenecarbonylimino-4,1-phenylenecarbonylimino)]bis-1,3,5-naphthalenetrisulfonic acid hexasodium salt
C49H36N6Na6O23S6
RRKORSRLXJQAGK-UHFFFAOYSA-N
131850945

Specifications

NF 279
202983-32-2
C49H36N6Na6O23S6
RRKORSRLXJQAGK-UHFFFAOYSA-N
sodium;8-[[4-[[4-[[4-[[4-[(4,6,8-trisulfonaphthalen-1-yl)carbamoyl]phenyl]carbamoyl]phenyl]carbamoylamino]benzoyl]amino]benzoyl]amino]naphthalene-1,3,5-trisulfonic acid
131850945
50mg
>98%
8,8'-[Carbonylbis(imino-4,1-phenylenecarbonylimino-4,1-phenylenecarbonylimino)]bis-1,3,5-naphthalenetrisulfonic acid hexasodium salt
C1=CC(=CC=C1C(=O)NC2=C3C(=CC(=CC3=C(C=C2)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O)NC(=O)C4=CC=C(C=C4)NC(=O)NC5=CC=C(C=C5)C(=O)NC6=CC=C(C=C6)C(=O)NC7=C8C(=CC(=CC8=C(C=C7)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na].[Na].[Na].[Na].[Na]
1407.145
1401.1
Videos