missing translation for 'onlineSavingsMsg'
Learn More

NF 110, Tocris Bioscience™

Potent P2X3 antagonist

Brand:  Tocris (Bio-Techne) 254810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13441444

  • £202.86 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

High affinity P2X3 receptor antagonist (Ki values are 36, 82 and 4144 nM for P2X3, P2X1 and P2X2 recombinant receptors respectively). Shows no activity at P2Y1, P2Y2 and P2Y11 receptors (IC50 > 10 μM). Potently inhibits α,β-meATP-evoked desensitizing currents in rat DRG neurons (IC50 = 527 nM). Shows antitumor activity against several tumor types.
Specifications

Chemical Identifiers

111150-22-2
1096.899
4,4',4,4'-carbonylbis imino-5,1,3-benzenetriylbis carbonylimino tetrakisbenzenesulfonic acid tetrasodium salt, benzenesulfonic acid,4,4',4,4'-carbonylbis imino-5,1,3-benzenetriylbis carbonylimino tetrakis-,tetrasodium salt 9ci, tetrasodium 4-3-3,5-bis 4-sulfonatophenyl carbamoyl phenyl carbamoyl amino-5-4-sulfonatophenyl carbamoyl benzamido benzenesulfonate
tetrasodium;4-[[3-[[3,5-bis[(4-sulfonatophenyl)carbamoyl]phenyl]carbamoylamino]-5-[(4-sulfonatophenyl)carbamoyl]benzoyl]amino]benzenesulfonate
C41H28N6Na4O17S4
AQJHZNCSXLBXMY-UHFFFAOYSA-J
16066783
C1=CC(=CC=C1NC(=O)C2=CC(=CC(=C2)NC(=O)NC3=CC(=CC(=C3)C(=O)NC4=CC=C(C=C4)S(=O)(=O)[O-])C(=O)NC5=CC=C(C=C5)S(=O)(=O)[O-])C(=O)NC6=CC=C(C=C6)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

10mg
111150-22-2
4,4',4,4'-carbonylbis imino-5,1,3-benzenetriylbis carbonylimino tetrakisbenzenesulfonic acid tetrasodium salt, benzenesulfonic acid,4,4',4,4'-carbonylbis imino-5,1,3-benzenetriylbis carbonylimino tetrakis-,tetrasodium salt 9ci, tetrasodium 4-3-3,5-bis 4-sulfonatophenyl carbamoyl phenyl carbamoyl amino-5-4-sulfonatophenyl carbamoyl benzamido benzenesulfonate
C1=CC(=CC=C1NC(=O)C2=CC(=CC(=C2)NC(=O)NC3=CC(=CC(=C3)C(=O)NC4=CC=C(C=C4)S(=O)(=O)[O-])C(=O)NC5=CC=C(C=C5)S(=O)(=O)[O-])C(=O)NC6=CC=C(C=C6)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
1096.899
1096.9
NF 110
C41H28N6Na4O17S4
AQJHZNCSXLBXMY-UHFFFAOYSA-J
tetrasodium;4-[[3-[[3,5-bis[(4-sulfonatophenyl)carbamoyl]phenyl]carbamoylamino]-5-[(4-sulfonatophenyl)carbamoyl]benzoyl]amino]benzenesulfonate
16066783
>98%
Videos
Documents