Learn More
NF 023, Tocris Bioscience™
Selective, competitive P2X1 antagonist
Brand:  Tocris (Bio-Techne) 124050
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13488943
  • £573.00 / 50mg
Estimated Shipment
02-07-2018
Add to basket
Description
Description
A subtype-selective, competitive and reversible P2X1 receptor antagonist. Displays IC50 values of 0.21, 28.9, > 50 and > 100 μM for human P2X1, P2X3, P2X2, and P2X4-mediated responses respectively. Selective over adrenoceptors, histamine and P2Y receptors. Also selectively inhibits the α-subunit of Go/i (EC50 ∽300 nM).
Specifications
Chemical Identifiers
NF 023
C35H20N4Na6O21S6
1162.86
8,8'-[carbonylbis(imino-3,1-phenylenecarbonylimino)]bis-1,3,5-naphthalene-trisulphonic acid, hexasodium salt
Specifications
NF 023
104869-31-0
C35H20N4Na6O21S6
8,8'-[carbonylbis(imino-3,1-phenylenecarbonylimino)]bis-1,3,5-naphthalene-trisulphonic acid, hexasodium salt
50mg
>98%
1162.86
Videos