missing translation for 'onlineSavingsMsg'
Learn More

New Pig Haz-Mat Pads

Stop the spread of acids and bases.

Brand:  New Pig MAT351

Code : Z4

Additional Details : Weight : 3.68000kg

 View more versions of this product

Product Code. 12439277

  • £65.03 / Pack of 100
Estimated Shipment: 15-04-2024
to see stock.
Add to basket

Description

Description

Stop the spread of acids and bases.

  • Chemical-resistant — won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Excellent for quickly diking and stopping the spread of chemical spills
  • Absorb and retain most acids, bases and unknown liquids
  • Can be incinerated after use to reduce waste

Specifications

Specifications

Pink
37.1 L
Hazmat Absorbent Pad
Dispenser
Heavyweight
13 in.
10 in.
8.1 lb.
12.54 oz.
Polypropylene
Quickly diking and stopping the spread of chemical spills, absorbs acids, bases, corrosives
13 L x 10 in. W (33.02 x 25.4 cm)
13 x 10 in. (33.02 x 25.4 cm)
33.02 cm
25.4 cm
3.7 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers