missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Neurotensin, Guinea pig

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J66243.MA

Code : NEW

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 15407529

  • £153.20 / 10mg


Estimated Shipment date
from Supplier 05-07-2018
Add to basket

Specifications

Specifications

1662.91
Guinea Pig
Solid
RUO
Neurotensin, guinea pig
HPLC
10mg
Glp-Leu-Tyr-Glu-Asn-Lys-Ser-Arg-Arg-Pro-Tyr-Ile-Leu
-30° to -10°C
Videos
SDS

For Research Use Only