missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Neurodazine
Induces neurogenesis in mature skeletal muscle cells
POA
Specifications
Purity | >99% |
---|---|
Product Type | Neurodazine |
Molecular Weight (g/mol) | 456.92 |
Inhibitors | Neurodazine |
Description
Induces neurogenesis of non-pluripotent C2C12 myoblasts and cells derived from mature human skeletal muscle. Upregulates the expression of neuron-specific markers in C2C12 cells.Specifications
>99% | |
Neurodazine | |
456.92 | |
Neurodazine |
For Research Use Only