Learn More
Tocris Bioscience™ Neurodazine
Induces neurogenesis in mature skeletal muscle cells
Brand:  Tocris Bioscience™ 365650
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443364
  • £445.92 / 50mg
Estimated Shipment
28-06-2019
Description
Description
Induces neurogenesis of non-pluripotent C2C12 myoblasts and cells derived from mature human skeletal muscle. Upregulates the expression of neuron-specific markers in C2C12 cells.
Specifications
Specifications
Neurodazine
>99
50mg
Neurodazine
456.92