missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Neurexophilin-1/NXPH-1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Neurexophilin-1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Neurexophilin-1 Polyclonal specifically detects Neurexophilin-1 in Rat samples. It is validated for Western Blot.Specifications
Neurexophilin-1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Rat | |
neurexophilin 1, neurexophilin-1, Neurexophilin1, NPH1, NXPH1 | |
NXPH1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q63366 | |
30010 | |
Mouse myeloma cell line NS0-derived recombinant rat Neurexophilin-1/NXPH-1 Ala22-Gly271 Accession # Q63366 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects rat Neurexophilin-1/NXPH-1 in direct ELISAs and Western blots. |
For Research Use Only