missing translation for 'onlineSavingsMsg'
Learn More

Neural/Glial Antigen 2 (NG2), clone: 9.2.27, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-6504-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15257717

  • £55.98 / 25µg


Estimated Shipment
27-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This 9.2.27 monoclonal antibody reacts with human neural/glial antigen 2, which is also known as melanoma chondroitin sulfate proteoglycan (MCSP). This antigen is composed of a 250kDa N-linked glycoprotein and a >450kDa proteoglycan. NG2 is present on the surface of >;90% of malignant melanomas in addition to some non-melanomic tumors. Moreover, NG2 is expressed on glioma cells, as well as on developing and adult oligodendrocyte precursor cells. Studies have demonstrated the involvement of this protein in tumor cell proliferation, adhesion, migration, and invasion. NG2 also functions as a coreceptor for alpha4beta1 integrin. Finally, expression of NG2 can be used as a prognostic marker for disorders such as acral letiginous melanoma and infantile acute myeloid leukemia.The 9.2.27 antibody has been reported to suppress melanoma tumor growth.Applications Reported: This 9.2.27 has been reported for use in flow cytometric analysis, immunoprecipitation, immunohistochemical staining of formalin-fixed paraffin embedded tissue (IHC-P), and immunocytochemical staining of fixed cells (ICC).Applications Tested: This 9.2.27 antibody has been tested by immunocytochemistry on methanol-fixed A375 cells at less than or equal to 10 ug/mL and by flow cytometric analysis on A375 cells at less than or equal to 0.125 ug/test. A test is defined as the amount (ug) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10e5 to 10e8 cells/test. It is recommended that this antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Neural / Glial Antigen 2 (NG2)
9.2.27
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
Immunoprecipitation
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Chondroitin Sulfate Proteoglycan, MCSP/melanoma chondroitin sulfate proteoglycan
IgG2a
RUO
Primary
Human
Videos