missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Netrin-G2a Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Netrin-G2a |
---|---|
Clone | 349724 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Description
Netrin-G2a Monoclonal specifically detects Netrin-G2a in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Netrin-G2a | |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rat | |
Mouse | |
NP_598007 | |
Ntng2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
349724 | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
84628 | |
Mouse myeloma cell line NS0-derived recombinant mouse Netrin-G2a Asp18-Arg504 Accession # NP_598007 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Netrin-G2a in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Netrin-G1a, rmNetrin-1, or rmNetrin-4 is observed. |
For Research Use Only