missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Netrin-G1a Rat anti-Mouse, Clone: 229722, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Netrin-G1a |
---|---|
Clone | 229722 |
Immunogen | S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Netrin-G1a His29-Gly513 Accession # Q8R4G0 |
Gene ID (Entrez) | 22854 |
Gene Accession No. | Q8R4G0 |
Description
Netrin-G1a Monoclonal specifically detects Netrin-G1a in Mouse samples. It is validated for Western Blot.Specifications
Netrin-G1a | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Netrin-G1a His29-Gly513 Accession # Q8R4G0 | |
Q8R4G0 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Rat | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
229722 | |
22854 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse Netrin-G1a in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant chicken (rch) Netrin-1, recombinant mouse Netrin-1, rchNetrin-2, or recombinant human Netrin-4 is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only