Learn More
Nestin Monoclonal Antibody (10C2), eFluor™ 570, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 41-9843-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Nestin is a Class VI intermediate filament abundantly produced in the developing central nervous system (CNS) and in early embryonic neuroepithelial stem cells. This cytoskeletal protein has been widely used as a predominant marker for stem/progenitor cells, glioma cells, and tumor endothelial cells. Also, it is a superior angiogenic marker to evaluate neovascularity of endothelial cells in tumors. These highly specific antibodies to human nestin can aid in characterizing progenitor cells differentiating into distinct lineages, in enhancing the therapeutic potential of human neural stem/progenitor cells in the treatment of CNS diseases or injury, and in identifying neuroepithelial tumor cells.
Nestin Monoclonal antibody specifically detects Nestin in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
Nestin | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P48681 | |
Nes | |
Affinity chromatography | |
RUO | |
10763 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
10C2 | |
eFluor 570 | |
Nes | |
AA166324; C78523; ESTM46; FLJ21841; intermediate filament protein; Nbla00170; Nes; Nestin | |
Mouse | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
For Research Use Only.