missing translation for 'onlineSavingsMsg'
Learn More

Nerol, 97%, Thermo Scientific Chemicals

Catalog Number 10649594
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
106-25-2
C10H18O
154.25
MFCD00063204
GLZPCOQZEFWAFX-YFHOEESVSA-N
nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol
643820
CHEBI:29452
(2Z)-3,7-dimethylocta-2,6-dien-1-ol
CC(C)=CCC\C(C)=C/CO
This item is not returnable. View return policy
106-25-2
C10H18O
154.25
MFCD00063204
GLZPCOQZEFWAFX-YFHOEESVSA-N
nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol
643820
CHEBI:29452
(2Z)-3,7-dimethylocta-2,6-dien-1-ol
CC(C)=CCC\C(C)=C/CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.8750g/mL
Boiling Point 225°C to 227°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CCH(CH2)2C(CH3)CHCH2OH
Refractive Index 1.4720 to 1.479
Quantity 500 mL
Beilstein 01, IV, 2276
Merck Index 15, 6560
Specific Gravity 0.875
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in absolute alcohol
Formula Weight 154.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Nerol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'