missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nepsilon-Boc-Nalpha-Fmoc-L-lysine pentafluorophenyl ester, 98%

Brand:  Alfa Aesar™ H61257.06

Code : NEW

Additional Details : CAS Number : 86060-98-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15476396

  • £208.82 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Nepsilon-Boc-Nalpha-Fmoc-L-lysine pentafluorophenyl ester
634.59
3582983
86060-98-2
MFCD00065661
Fmoc-Lys(Boc)-OPfp

Specifications

Nepsilon-Boc-Nalpha-Fmoc-L-lysine pentafluorophenyl ester
Solid
634.59
3582983
5g
86060-98-2
98%
MFCD00065661
Fmoc-Lys(Boc)-OPfp
Videos
SDS