missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nepsilon-Boc-D-lysine, 98%

Brand:  Alfa Aesar™ H62736.03

Code : NEW

Additional Details : CAS Number : 31202-69-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449218

  • £74.20 / 1g


Estimated Shipment date
from Supplier 22-02-2018
Add to basket

Specifications

Chemical Identifiers

Nepsilon-Boc-D-lysine
246.3
H-D-Lys(Boc)-OH; (R)-2-Amino-6-(Boc-amino)hexanoic acid
31202-69-4
MFCD00076958

Specifications

Nepsilon-Boc-D-lysine
Powder
246.3
H-D-Lys(Boc)-OH; (R)-2-Amino-6-(Boc-amino)hexanoic acid
1g
31202-69-4
98%
MFCD00076958
White
Videos
SDS