Learn More
Alfa Aesar™ Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester, 95%
Brand: Alfa Aesar™ H63470.14
Code : NEW
Additional Details : CAS Number : 34404-36-9 Weight : 0.02500kg
Quantity | 25g |
---|
Description
SolubilitySlightly soluble in water.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester | |
477.51 | |
1559393 |
34404-36-9 | |
MFCD00037915 | |
Boc-Lys(Z)-OSu |
Specifications
Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester | |
Powder | |
477.51 | |
1559393 | |
White | |
25g |
34404-36-9 | |
95% | |
MFCD00037915 | |
Boc-Lys(Z)-OSu | |
108° to 117°C |