missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester, 95%

Brand:  Alfa Aesar™ H63470.06

Code : NEW

Additional Details : CAS Number : 34404-36-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15458326

  • £56.80 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Description

Description

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester
477.51
1559393
34404-36-9
MFCD00037915
Boc-Lys(Z)-OSu

Specifications

Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester
Powder
477.51
1559393
White
5g
34404-36-9
95%
MFCD00037915
Boc-Lys(Z)-OSu
108° to 117°C
Videos
SDS