missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-L-lysine, 95%, Thermo Scientific Chemicals

Catalog Number 15490629
23.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
146982-27-6
C25H28N2O6
452.51
MFCD00190872
OJBNDXHENJDCBA-ANBDAQEENA-N
fmoc-lys alloc-oh, fmoc-lys aloc-oh, n-fmoc-n'-allyoxycarbonyl-l-lysine, s-2-9h-ffuoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, fmoc-lys alloc, aloc-lys fmoc-oh, pubchem13153, fmoc-l-lys alloc-oh, n, a-fmoc-n, a-alloc-l-lysine
11123257
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(prop-2-enoxycarbonylamino)hexanoic acid
OC(=O)[C@H](CCCCNC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
146982-27-6
C25H28N2O6
452.51
MFCD00190872
OJBNDXHENJDCBA-ANBDAQEENA-N
fmoc-lys alloc-oh, fmoc-lys aloc-oh, n-fmoc-n'-allyoxycarbonyl-l-lysine, s-2-9h-ffuoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, fmoc-lys alloc, aloc-lys fmoc-oh, pubchem13153, fmoc-l-lys alloc-oh, n, a-fmoc-n, a-alloc-l-lysine
11123257
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(prop-2-enoxycarbonylamino)hexanoic acid
OC(=O)[C@H](CCCCNC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Beilstein 5893081
Formula Weight 452.5
Percent Purity 95%
Quantity 1 g
Physical Form Solid
Chemical Name or Material Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-L-lysine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'