missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-D-lysine, 95%

Brand:  Alfa Aesar™ H66861.03

Code : NEW

Additional Details : CAS Number : 214750-75-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402839

  • £23.41 / 1g

Estimated Shipment: 26-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

214750-75-1
452.507
OJBNDXHENJDCBA-JOCHJYFZSA-N
51340505
C=CCOC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C25H28N2O6
MFCD00798639
fmoc-d-lys alloc-oh, fmoc-d-lys alloc-oh hplc, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-allyl-oxycarbonyl-d-lysine, n~2~-9h-fluoren-9-yl methoxy carbonyl-n~6~-prop-2-en-1-yl oxy carbonyl-d-lysine, fmoc-d-lys aloc-oh, n-fmoc-n'-allyloxycarbonyl-d-lysine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, ambotzfaa1329, n, a-alloc-n, a-fmoc-d-lysine, a-fmoc-n, a-alloc-d-lysine
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(prop-2-enoxycarbonylamino)hexanoic acid

Specifications

Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-D-lysine
C25H28N2O6
fmoc-d-lys alloc-oh, fmoc-d-lys alloc-oh hplc, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-allyl-oxycarbonyl-d-lysine, n~2~-9h-fluoren-9-yl methoxy carbonyl-n~6~-prop-2-en-1-yl oxy carbonyl-d-lysine, fmoc-d-lys aloc-oh, n-fmoc-n'-allyloxycarbonyl-d-lysine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, ambotzfaa1329, n, a-alloc-n, a-fmoc-d-lysine, a-fmoc-n, a-alloc-d-lysine
C=CCOC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
452.507
452.51
95%
1g
214750-75-1
MFCD00798639
OJBNDXHENJDCBA-JOCHJYFZSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(prop-2-enoxycarbonylamino)hexanoic acid
51340505
Powder
White
Videos
SDS Documents
Certificates