missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-D-lysine, 95%

Brand:  Alfa Aesar™ H66861.06

Code : NEW

Additional Details : CAS Number : 214750-75-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15412839

  • £84.10 / 5g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-D-lysine
452.51
Fmoc-D-Lys(Aloc)-OH
214750-75-1
MFCD00798639

Specifications

Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-D-lysine
Powder
452.51
Fmoc-D-Lys(Aloc)-OH
5g
214750-75-1
95%
MFCD00798639
White
Videos
SDS