missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals

Catalog Number 15489508
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
148083-64-1
C27H26N4O8
534.53
MFCD00273454
OENCMORJQAUAAJ-FQKVKQEKNA-N
fmoc-lys dnp-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-2,4-dinitrophenyl amino hexanoic acid, fmoc-l-lys dnp-oh, n-fmoc-n'-2,4-dinitrophenyl-l-lysine, fmoc-lys dnp, 2s-6-2,4-dinitrophenyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-2,4-dinitrophenyl-l-lysine
44593781
6-[(2,4-dinitrophenyl)amino]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
OC(=O)[C@H](CCCCNC1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
This item is not returnable. View return policy
148083-64-1
C27H26N4O8
534.53
MFCD00273454
OENCMORJQAUAAJ-FQKVKQEKNA-N
fmoc-lys dnp-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-2,4-dinitrophenyl amino hexanoic acid, fmoc-l-lys dnp-oh, n-fmoc-n'-2,4-dinitrophenyl-l-lysine, fmoc-lys dnp, 2s-6-2,4-dinitrophenyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-2,4-dinitrophenyl-l-lysine
44593781
6-[(2,4-dinitrophenyl)amino]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
OC(=O)[C@H](CCCCNC1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Formula Weight 534.52
Percent Purity 98%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'