missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine, 98%

Brand:  Alfa Aesar™ H63050.06

Code : NEW

Additional Details : CAS Number : 148083-64-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15479508

  • £218.10 / 5g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine
534.52
Fmoc-Lys(Dnp)-OH
148083-64-1
MFCD00273454

Specifications

Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine
Solid
534.52
Fmoc-Lys(Dnp)-OH
148083-64-1
98%
MFCD00273454
5g
Videos
SDS