Learn More
Neomycin sulfate hydrate, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Neomycin sulfate is an aminoglycoside antibiotic and calcium channel protein inhibitor. Neomycin sulfate also binds to prokaryotic ribosomes inhibiting translation and is effective against gram-positive and gram-negative bacteria. Neomycin sulfate inhibits Phospholipase C via binding to inositol phospholipids. It also inhibits phosphatidylcholine-PLD activity and induces Ca2+ mobilization and PLA2 activation in human platelets. Neomycin sulfate inhibits DNase I induced DNA degradation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Neomycin sulfate is an aminoglycoside antibiotic and calcium channel protein inhibitor. Neomycin sulfate also binds to prokaryotic ribosomes inhibiting translation and is effective against gram-positive and gram-negative bacteria. Neomycin sulfate inhibits Phospholipase C via binding to inositol phospholipids. It also inhibits phosphatidylcholine-PLD activity and induces Ca2+ mobilization and PLA2 activation in human platelets. Neomycin sulfate inhibits DNase I induced DNA degradation.
Solubility
Soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Type | Neomycin Sulfate |
Physical Form | Powder |
Merck Index | 14,6454 |
Solubility Information | Soluble in water |
IUPAC Name | (2S,3R,4S,5S,6R)-5-amino-2-(aminomethyl)-6-[(1R,2R,3S,4R,6R)-4,6-diamino-2-[(2R,3S,4R,5S)-4-[(2S,3R,4S,5R,6S)-3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy-3-hydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-3-hydroxycyclohexyl]oxyoxane-3,4-diol;sulfuric acid |
Formula Weight | 908.87 (Anhydrous) |
Color | White |
Quantity | 100 g |
Chemical Name or Material | Neomycin sulfate hydrate |
RUO – Research Use Only