missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Neogenin Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Neogenin |
---|---|
Clone | 221519 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Neogenin Monoclonal specifically detects Neogenin in Mouse samples. It is validated for Western Blot.Specifications
Neogenin | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
NP_032710 | |
4756 | |
Mouse myeloma cell line NS0-derived recombinant mouse Neogenin Ala42-Ile1033 (Asp442-Leu461 del) Accession # NP_032710 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Neogenin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse DCC is observed. |
221519 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HsT17534, IGDCC2DKFZp547B146, Immunoglobulin superfamily DCC subclass member 2, NEO, NEO1, neogenin, neogenin (chicken) homolog 1, neogenin 1, neogenin homolog 1, NGN, NGNDKFZp547A066 | |
NEO1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only