missing translation for 'onlineSavingsMsg'
Learn More

Neodymium ingot, 99.1% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048201
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
7440-00-8
Nd
144.24
MFCD00011130
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
23934
CHEBI:33372
neodymium
[Nd]
This item is not returnable. View return policy
7440-00-8
Nd
144.24
MFCD00011130
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
23934
CHEBI:33372
neodymium
[Nd]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under mineral oil
TRUSTED_SUSTAINABILITY
Color Gray-Silver
Quantity 25 g
Percent Purity 99.1%
Form Ingot
Assay (metals basis)
Chemical Name or Material Neodymium ingot

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'