missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Neodymium(III) sulfate octahydrate, 99.999% (REO)

Brand:  Alfa Aesar™ 041855.09

Code : NEW

Additional Details : CAS Number : 13477-91-3 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11384817

  • £78.60 / 10g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

Neodymium(III) sulfate octahydrate
720.78 (576.66 Anhydrous)
14,6450
13477-91-3
MFCD00149803

Specifications

Neodymium(III) sulfate octahydrate
Crystalline
(REO)
MFCD00149803
2.85g/mL
1176°C
13477-91-3
99.999%
720.78 (576.66 Anhydrous)
14,6450
Odorless
10g
Videos
SDS